Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin B8/Proteinase Inhibitor 8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4158
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4158 100 μg
AF4158SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4158 Supplier R&D Systems Supplier No. AF4158
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin B8/Proteinase Inhibitor 8 Polyclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Serpin B8/Proteinase Inhibitor 8
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P50452
Gene Alias CAP2, CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, NK10, Peptidase inhibitor 8, PI8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8
Gene Symbols SERPINB8
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin B8/Proteinase Inhibitor 8 Asp2-Pro374 Accession # P50452
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5271
Test Specificity Detects human Serpin B8/Proteinase Inhibitor 8 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Serpin B8 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin B6, rhSerpin B9, and rhSerpin A5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.