Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin C1/Antithrombin-III Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $561.00
Specifications
Antigen | Serpin C1/Antithrombin-III |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.6 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Description
Serpin C1/Antithrombin-III Polyclonal specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Serpin C1/Antithrombin-III | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.6 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Antithrombin-III, AT3antithrombin-III, ATIII, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
SERPINC1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
P01008 | |
462 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin C1/Antithrombin-III His33-Lys464 Accession # P01008 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Serpin C1/Antithrombin-III in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse (rm) Serpin C1 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rhSerpin F2, rmSerpin D1 and rhPEDF is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title