Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin D1/Heparin Cofactor II Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | Serpin D1/Heparin Cofactor II |
---|---|
Clone | 373008 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Serpin D1/Heparin Cofactor II | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P05546 | |
3053 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin D1/Heparin Cofactor II Gly20-Arg498 Accession # P05546 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serpin D1/Heparin Cofactor II in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A3, A4, B6, B8, C1, I1, I2, recombinant mouse Serpin A1, A6, D1, E1, E2, F1, or F2 is observed. |
373008 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HC2, HC2LS2, HCF2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, LS2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1 | |
SERPIND1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title