Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin F1/PEDF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1177100
This item is not returnable.
View return policy
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Serpin F1/PEDF | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P36955 | |
SERPINF1 | |
E. coli-derived recombinant human Serpin F1, Mouse myeloma cell line NS0-derived recombinant human Serpin F1, and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin F1 Asp44-Pro418 Accession # P36955 | |
100 μg | |
Primary | |
Detects human Serpin F1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Serpin F1 is observed. | |
Human | |
Supernatant |
Western Blot, Immunohistochemistry | |
187003 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5176 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction