Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serpin F1/PEDF Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1177
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1177 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1177 Supplier R&D Systems Supplier No. BAF1177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin F1/PEDF Polyclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot.

Specifications

Antigen Serpin F1/PEDF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Alias Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1
Gene Symbols SERPINF1
Host Species Goat
Immunogen recombinant human Serpin F1/PEDF
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5176
Test Specificity Detects human Serpin F1/PEDF in Western blots. In this format, approximately 40% cross-reactivity with recombinant mouse Serpin F1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.