Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serpin F2/alpha 2-Antiplasmin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $596.00
Specifications
Antigen | Serpin F2/alpha 2-Antiplasmin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.2 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Description
Serpin F2/alpha 2-Antiplasmin Polyclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Serpin F2/alpha 2-Antiplasmin | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.2 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
SERPINF2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
P08697 | |
5345 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2 -Antiplasmin Met28-Lys491 Accession # P08697 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Serpin F2/alpha 2 -Antiplasmin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Serpin F2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rmSerpin C1, and rmSerpin D1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title