Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serum Albumin Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1455R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1455R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1455R100 Supplier R&D Systems Supplier No. IC1455R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Albumin
Applications Flow Cytometry
Classification Monoclonal
Clone 188835
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Mouse
Immunogen Human Serum Albumin
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity Detects human Serum Albumin in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.