Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serum Albumin Antibody (2B12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178553
Description
Human Serum Albumin Monoclonal specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.Specifications
Human Serum Albumin | |
Monoclonal | |
1 mg/mL | |
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
2B12 | |
Unconjugated | |
P02768 | |
ALB | |
Human Serum Albumin | |
100 μg | |
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction