Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Serum Albumin Native Protein
SDP

Catalog No. N19959810U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N19959810U 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N19959810U Supplier Novus Biologicals™ Supplier No. NBP1995981000UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page

Native Serum Albumin

The Human Serum Albumin Native Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 250 mg/mL
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 213
Molecular Weight (g/mol) 66.4 kDa
Quantity 1000 μg
Source Human serum
Storage Requirements Store at 4°C.
Regulatory Status RUO
Gene Symbol ALB
Specific Reactivity Human Serum Albumin
Conjugate Unconjugated
Species Human
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.