Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serum Albumin PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1455P
This item is not returnable.
View return policy
Description
Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Albumin | |
Monoclonal | |
R-PE | |
ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Serum Albumin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
188835 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
ALB | |
Human Serum Albumin | |
100 Tests | |
213 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction