Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serum Amyloid A1/A2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Serum Amyloid A1/A2 |
---|---|
Clone | 865504 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB30196SP
![]() |
R&D Systems
MAB30196SP |
25 μg |
Each for $200.50
|
|
|||||
MAB30196100
![]() |
R&D Systems
MAB30196100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for ELISA.Specifications
Serum Amyloid A1/A2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
6288 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
865504 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P0DJI8 | |
E. coli-derived recombinant human Serum Amyloid A1 Arg19-Tyr122 Accession # P0DJI8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serum Amyloid A1 and Serum Amyloid A2 in direct ELISAs. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title