Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Serum Amyloid A1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Serum Amyloid A1 |
---|---|
Clone | 370820 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Serum Amyloid A1 Monoclonal specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot.Specifications
Serum Amyloid A1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P02735 | |
6288 | |
E. coli-derived recombinant human SAA1 Arg19-Tyr122 Accession # P02735 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serum Amyloid A1 (SAA1) in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse SAA1 and no cross-reactivity with recombinant human (rh) SAA4 is observed. In direct ELISAs, no cross-reactivity with rhSAA2 is observed. |
370820 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC111216, PIG4, SAA, SAA1, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
SAA1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title