Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serum Amyloid A4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3856SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3856SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3856SP Supplier R&D Systems Supplier No. AF3856SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Serum Amyloid A4 Polyclonal specifically detects Serum Amyloid A4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Serum Amyloid A4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35542
Gene Alias C-SAA, C-SAACSAAConstitutively expressed serum amyloid A protein, SAA4, SAA5, serum amyloid A-4 protein, serum amyloid A4, constitutive
Gene Symbols SAA4
Host Species Goat
Immunogen E. coli-derived recombinant human Serum Amyloid A4 Glu19-Tyr130 Accession # P35542
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6291
Test Specificity Detects human Serum Amyloid A4 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Serum Amyloid A4 is observed and less than 5% cross-reactivity with recombinant human (rh) Serum Amyloid A1 and rhSerum Amyloid A2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.