Learn More
Human Serum Polyclonal Antibody, HRP, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $612.54
Specifications
| Antigen | Human Serum |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1449
![]()
|
Rockland Immunochemicals
20943010100 |
100 μg |
Each for $612.54
|
|
|||||
50-199-1450
![]()
|
Rockland Immunochemicals
2094301S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Human serum is an IgG fraction antibody purified from polyspecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase and anti-Rabbit Serum.
Human serum is obtained by pooling the liquid portion of coagulated human whole blood. Rabbit Anti-Human Serum Antibody is useful for researchers interested in serum component proteins.Specifications
| Human Serum | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| HRP | |
| Rabbit | |
| Human | |
| Human serum proteins | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and no preservative; pH 7.2 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.