Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Serum TIM-1/KIM-1/HAVCR Quantikine ELISA Kit
SDP

Catalog No. DSKM100
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DSKM100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DSKM100 Supplier R&D Systems™ Supplier No. DSKM100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NS0-expressed recombinant human TIM-1

For the determination of natural and recombinant TIM-1 in humans. R&D Systems Human Serum TIM-1/KIM-1/HAVCR Quantikine™ ELISA Kit is a 4.5 hour solid-phase ELISA designed to measure TIM-1 in human cell culture supernates, serum and plasma.

The Human Serum TIM-1/KIM-1/HAVCR Quantikine ELISA Kit quantifies human TIM-1/KIM-1/HAVCR in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Description CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Product Type ELISA Kit
Quantity 1 Kit
Species Human
Assay Sensitivity 3.63 pg/mL
Assay Range 10.90 to 700 pg/mL
Cross Reactivity <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested
For Use With (Application) ELISA
Gene ID (Entrez) 26762
Sample Type Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Sample Volume 50 μL (Serum), 50 μL (Cell Culture Supernates), 50 μL (Heparin plasma), 50 μL (EDTA plasma)
Test Time 4.5 hr.
Synonym CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Kit Contents NS0-expressed recombinant human TIM-1
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.