Learn More
Human SF20/MYDGF Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | SF20/MYDGF |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1009420 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q969H8 |
| Gene Alias | C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL-25, IL25IL27w, IL27, IL-27, interleukin 25, interleukin 27 working designation, Interleukin-25, Ly6elg, MYDGF, R33729_1, SF20hypothetical protein LOC56005, stromal cell-derived growth factor, Stromal cell-derived growth factor SF20 |
| Gene Symbols | MYDGF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.