Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SF20/MYDGF Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1147V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1147V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1147V100 Supplier R&D Systems Supplier No. IC1147V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen SF20/MYDGF
Applications Flow Cytometry
Classification Monoclonal
Clone 1009420
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q969H8
Gene Alias C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL-25, IL25IL27w, IL27, IL-27, interleukin 25, interleukin 27 working designation, Interleukin-25, Ly6elg, MYDGF, R33729_1, SF20hypothetical protein LOC56005, stromal cell-derived growth factor, Stromal cell-derived growth factor SF20
Gene Symbols MYDGF
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SF20, Met1-Leu173, Accession # Q969H8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 56005
Test Specificity Detects human SF20 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.