Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SF3A1 Full-length ORF (CAK54605.1, 1 a.a. - 793 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961613 Shop All Abnova Corporation Products
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes subunit 1 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 1 belongs to the SURP protein family; named for the SURP (also called SWAP or Suppressor-of-White-APricot) motifs that are thought to mediate RNA binding. Subunit 1 has tandemly repeated SURP motifs in its amino-terminal half while its carboxy-terminal half contains a proline-rich region and a ubiquitin-like domain. Binding studies with truncated subunit 1 derivatives demonstrated that the two SURP motifs are necessary for binding to subunit 3 while contacts with subunit 2 may occur through sequences carboxy-terminal to the SURP motifs. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MPAGPVQAVPPPPPVPTEPKQPTEEEASSKEDSAPSKPVVGIIYPPPEVRNIVDKTASFVARNGPEFEARIRQNEINNPKFNFLNPNDPYHAYYRHKVSEFKEGKAQEPSAAIPKVMQQQQQTTQQQLPQKVQAQVIQETIVPKEPPPEFEFIADPPSISAFDLDVVKLTAQFVARNGRQFLTQLMQKEQRNYQFDFLRPQHSLFNYFTKLVEQYTKILIPPKGLFSKLKKEAENPREVLDQVCYRVEWAKFQERERKKEEEEKEKERVAYAQIDWHDFVVVETVDFQPNEQGNFPPPTTPEELGARILIQERYEKFGESEEVEMEVESDEEDDKQEKAEEPPSQLDQDTQVQDMDEGSDDEEEGQKVPPPPETPMPPPLPPTPDQVIVRKDYDPKASKPLPPAPAPDEYLVSPITGEKIPASKMQEHMRIGLLDPRWLEQRDRSIREKQSDDEVYAPGLDIESSLKQLAERRTDIFGVEETAIGKKIGEEEIQKPEEKVTWDGHSGSMARTQQAAQANITLQEQIEAIHKAKGLVPEDDTKEKIGPSKPNEIPQQPPPPSSATNIPSSAPPITSVPRPPTMPPPVRTTVVSAVPVMPRPPMASVVRLPPGSVIAPMPPIIHAPRINVVPMPPSAPPIMAPRPPPMIVPTAFVPAPPVAPVPAPAPMPPVHPPPPMEDEPTSKKLKTEDSLMPEEEFLRRNKGPVSIKVQVPNMQDKTEWKLNGQVLVFTLPLTDQVSVIKVKIHEATGMPAGKQKLQYEGIFIKDSNSLAYYNMANGAVIHLALKERGGRKK

Specifications

Accession Number CAK54605.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10291
Molecular Weight (g/mol) 113.63kDa
Name SF3A1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MPAGPVQAVPPPPPVPTEPKQPTEEEASSKEDSAPSKPVVGIIYPPPEVRNIVDKTASFVARNGPEFEARIRQNEINNPKFNFLNPNDPYHAYYRHKVSEFKEGKAQEPSAAIPKVMQQQQQTTQQQLPQKVQAQVIQETIVPKEPPPEFEFIADPPSISAFDLDVVKLTAQFVARNGRQFLTQLMQKEQRNYQFDFLRPQHSLFNYFTKLVEQYTKILIPPKGLFSKLKKEAENPREVLDQVCYRVEWAKFQERERKKEEEEKEKERVAYAQIDWHDFVVVETVDFQPNEQGNFPPPTTPEELGARILIQERYEKFGESEEVEMEVESDEEDDKQEKAEEPPSQLDQDTQVQDMDEGSDDEEEGQKVPPPPETPMPPPLPPTPDQVIVRKDYDPKASKPLPPAPAPDEYLVSPITGEKIPASKMQEHMRIGLLDPRWLEQRDRSIREKQSDDEVYAPGLDIESSLKQLAERRTDIFGVEETAIGKKIGEEEIQKPEEKVTWDGHSGSMARTQQAAQANITLQEQIEAIHKAKGLVPEDDTKEKIGPSKPNEIPQQPPPPSSATNIPSSAPPITSVPRPPTMPPPVRTTVVSAVPVMPRPPMASVVRLPPGSVIAPMPPIIHAPRINVVPMPPSAPPIMAPRPPPMIVPTAFVPAPPVAPVPAPAPMPPVHPPPPMEDEPTSKKLKTEDSLMPEEEFLRRNKGPVSIKVQVPNMQDKTEWKLNGQVLVFTLPLTDQVSVIKVKIHEATGMPAGKQKLQYEGIFIKDSNSLAYYNMANGAVIHLALKERGGRKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PRP21/PRPF21/SAP114/SF3A120
Common Name SF3A1
Gene Symbol SF3A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.