Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human sFRP-1 ELISA Kit

Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Description
Human sFRP-1 quantitates human sFRP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human sFRP-1.
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.

Specifications
Specifications
Accession Number | 2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; Sfrp1; sFRP-1 |
Assay Range | 0.78 to 50 ng/mL |
Assay Sensitivity | 0.78 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 6422 |
Gene Symbol | SFRP1 |
Interassay CV | <12% |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction