Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human sFRP-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6838
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6838 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6838 Supplier R&D Systems Supplier No. MAB6838
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

sFRP-2 Monoclonal specifically detects sFRP-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen sFRP-2
Applications Western Blot
Classification Monoclonal
Clone 755644
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96HF1
Gene Alias FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2
Gene Symbols SFRP2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human sFRP-2 Leu25-Cys295 Accession # Q96HF1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6423
Test Specificity Detects human sFRP-2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) sFRP-2 is observed and no cross-reactivity with recombinant human (rh) sFRP-1 or rhsFRP-5 is observed. In Western blots, no cross-reactivity with rhsFRP-1, -4, -5, or rmsFRP-2 is observed. 100% cross-reactivity with rhsFRP-3 is observed under non-reducing conditions only.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.