Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SGPL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5535 Supplier R&D Systems Supplier No. AF5535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

SGPL1 Polyclonal specifically detects SGPL1 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen SGPL1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL
Gene Accession No. O95470
Gene Alias EC 4.1.2.27, FLJ13811, hSPL, KIAA1252, S1PL, Sphingosine-1-phosphate aldolase, sphingosine-1-phosphate lyase 1, SPL, SPLSPL 1, SP-lyase, SP-lyase 1
Gene Symbols SGPL1
Host Species Goat
Immunogen E. coli-derived recombinant human SGPL1 Lys342-His568 Accession # O95470
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8879
Test Specificity Detects human SGPL1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.