Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SH2D1A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7440SP
This item is not returnable.
View return policy
Description
SH2D1A Polyclonal specifically detects SH2D1A in Human samples. It is validated for Western Blot.Specifications
SH2D1A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DSHP, DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, LYP, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLP, XLPD, XLPDFLJ18687, XLPSH2 domain protein 1A | |
Sheep | |
Affinity Purified | |
RUO | |
4068 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
O60880 | |
SH2D1A | |
E. coli-derived recombinant human SH2D1A Met1-Pro128 Accession # O60880 | |
25 μg | |
Primary | |
Detects human SH2D1A in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction