Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SH3BGRL2 (NP_113657, 1 a.a. - 107 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89961789
Click to view available options
Quantity:
100 μg

Used for SDS-PAGE

Sequence: MGSSHHHHHHSSGLVPRGSHMGSMVIRVFIASSSGFVAIKKKQQDVVRFLEANKIEFEEVDITMSEEQRQWMYKNVPPEKKPTQGNPLPPQIFNGDRYCGDYDSFFESKESNTVFSFLGLKPRLASKAEP

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 83699
Molecular Weight (g/mol) 14.7kDa
Name SH3BGRL2 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed.
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGSMVIRVFIASSSGFVAIKKKQQDVVRFLEANKIEFEEVDITMSEEQRQWMYKNVPPEKKPTQGNPLPPQIFNGDRYCGDYDSFFESKESNTVFSFLGLKPRLASKAEP
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ90025
Common Name SH3BGRL2
Gene Symbol SH3BGRL2
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS - PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.