Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SHANK2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7035SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7035SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7035SP Supplier R&D Systems Supplier No. AF7035SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SHANK2 Polyclonal specifically detects SHANK2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SHANK2
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UPX8
Gene Alias AUTS17, cortactin binding protein 1, cortactin SH3 domain-binding protein, Cortactin-binding protein 1, CortBP1, CORTBP1CTTNBP1, CTTNBP1, GKAP/SAPAP interacting protein, KIAA1022, proline-rich synapse associated protein 1, Proline-rich synapse-associated protein 1, PROSAP1, SH3 and multiple ankyrin repeat domains 2, SH3 and multiple ankyrin repeat domains protein 2, SHANK, Shank2, SPANK-3
Gene Symbols SHANK2
Host Species Sheep
Immunogen E. coli-derived recombinant human SHANK2 Lys849-Thr1029 Accession # Q9UPX8
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22941
Test Specificity Detects human SHANK2 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.