Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SHB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7036
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7036 Supplier R&D Systems Supplier No. AF7036
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SHB Polyclonal specifically detects SHB in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SHB
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q15464
Gene Alias bA3J10.2, RP11-3J10.8, SH2 domain-containing adapter protein B, SHB (Src homology 2 domain containing) adaptor protein B, SHB adaptor protein (a Src homology 2 protein), Src homology 2 domain containing adaptor protein B
Gene Symbols SHB
Host Species Goat
Immunogen E. coli-derived recombinant human SHB Ala2-Phe128 Accession # Q15464
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6461
Test Specificity Detects human SHB in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.