Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SHBG Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $502.50

Specifications

Antigen SHBG
Clone 336729
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB26562SP
GREENER_CHOICE
View Documents
R&D Systems
MAB26562SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB26562100
GREENER_CHOICE
View Documents
R&D Systems
MAB26562100
100 μg
Each for $502.50
Only null left
Add to Cart
 
Description

Description

SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for ELISA.
Specifications

Specifications

SHBG
ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
Mouse myeloma cell line NS0-derived recombinant human SHBG Leu30-His402 Accession # P04278
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human SHBG in direct ELISAs.
336729
ELISA
Unconjugated
Mouse
Human
P04278
6462
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

RUO

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.