Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SHPRH Partial ORF (NP_775105, 1560 a.a. - 1659 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961883
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

SHPRH is a ubiquitously expressed protein that contains motifs characteristics of several DNA repair proteins, transcription factors, and helicases. SHPRH is a functional homolog of S. cerevisiae RAD5 (Unk et al., 2006 [PubMed 17108083]).[supplied by OMIM]

Sequence: AQISRVKTFQENLSAFKRDPQINILLLPLHTGSNGLTIIEATHVLLVEPILNPAHELQAIGRVHRIGQTKPTIVHRFLIKATIEERMQAMLKTAERSIYI

Specifications

Accession Number NP_775105
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 257218
Molecular Weight (g/mol) 36.74kDa
Name SHPRH (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen AQISRVKTFQENLSAFKRDPQINILLLPLHTGSNGLTIIEATHVLLVEPILNPAHELQAIGRVHRIGQTKPTIVHRFLIKATIEERMQAMLKTAERSIYI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ27258/FLJ37625/FLJ45012/FLJ90837/KIAA2023/MGC134886/bA545I5.2
Common Name SHPRH
Gene Symbol SHPRH
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.