Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIGIRR APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB990A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB990A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB990A Supplier R&D Systems Supplier No. FAB990A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIGIRR Monoclonal specifically detects SIGIRR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SIGIRR
Applications Flow Cytometry
Classification Monoclonal
Clone 162201
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q6IA17
Gene Alias IL-1R8, single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8, TIR8MGC110992, Toll/interleukin-1 receptor 8
Gene Symbols SIGIRR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIGIRR Met1-His118 Accession # Q6IA17
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59307
Test Specificity Detects human SIGIRR in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) IL-1 RI, rhIL-1 R3, rhIL-1 R8, rhIL-1 RII, rhIL-1 R6, rhIL-1 R4, rhIL-18, rhIL-1 R7, rhIL-9 R9, or recombinant mouse SIGIRR.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.