Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-10 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2130
This item is not returnable.
View return policy
Description
Siglec-10 Polyclonal specifically detects Siglec-10 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
Siglec-10 | |
Polyclonal | |
Unconjugated | |
Q96LC7 | |
SIGLEC10 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-10 Met17-Thr546 Accession # Q96LC7 | |
100 μg | |
Primary | |
Detects human Siglec-10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Siglec-5 is observed and less than 1% cross-reactivity with rhSiglec-2, rhSiglec-3, rhSiglec-7, rhSiglec-9, and recombinant mouse Siglec-F is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, Siglec10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7 | |
Goat | |
Affinity Purified | |
RUO | |
89790 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction