Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-11 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3258P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3258P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3258P Supplier R&D Systems Supplier No. FAB3258P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Siglec-11 Polyclonal specifically detects Siglec-11 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-11
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q96RL6
Gene Alias sialic acid binding Ig-like lectin 11, sialic acid-binding Ig-like lectin 11, Sialic acid-binding lectin 11, siglec-11, Siglec11
Gene Symbols SIGLEC11
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Siglec-11 Asn17-His543 (Glu84Ala, Lys145Gln) Accession # Q96RL6
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114132
Test Specificity Detects human Siglec-11 in direct ELISAs and Western blots. In direct ELISA and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Siglec-10 is observed and less than 5% cross-reactivity with rhSiglec-2, -3, -5, -6, -7, -9 and recombinant mouse Siglec-F is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.