Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-16 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB68192G100UG
This item is not returnable.
View return policy
Description
Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow CytometrySpecifications
Siglec-16 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Siglec16, Siglec-P16, SIGLECP16 | |
Human Siglec-16 peptide, Accession # A6NMB1 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
706030 | |
Flow Cytometry 0.25-1 ug/106 cells | |
A6NMB1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
400709 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction