Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB68191SP
This item is not returnable.
View return policy
Description
Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Siglec-16 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Siglec16, Siglec-P16, SIGLECP16 | |
Human Siglec-16 peptide, Accession # A6NMB1 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
706045 | |
Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
A6NMB1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
400709 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction