Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-2/CD22 LlaMABodyTM Bivalent VHH HuIgG2 Fusion Antibody, R&D Systems™

Llama Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | Siglec-2/CD22 |
---|---|
Clone | L007.2.5H |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
LMAB107321S
![]() |
R&D Systems
LMAB107321SP |
25 μg |
Each for $200.50
|
|
|||||
LMAB1073211
![]() |
R&D Systems
LMAB107321100 |
100 μg |
Each for $410.00
|
|
|||||
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for ImmunocytochemistrySpecifications
Siglec-2/CD22 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
Chinese Hamster Ovary cell line CHO-derived human Siglec-2/CD22 as immunogen for bivalent Llama VHH-Human IgG2 Fusion Antibody, Asp20-Arg687, Accession # CAA42006.1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
L007.2.5H | |
Immunocytochemistry | |
Unconjugated | |
Llama | |
Human | |
CAA42006.1 | |
933 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title