Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-2/CD22 LlaMABodyTM VHH Antibody, R&D Systems™

Llama Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | Siglec-2/CD22 |
|---|---|
| Clone | L007.2.5LL |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
LMAB11048SP
![]() |
R&D Systems
LMAB11048SP |
25 μg |
Each for $200.50
|
|
|||||
LMAB1104810
![]() |
R&D Systems
LMAB11048100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for ImmunocytochemistrySpecifications
| Siglec-2/CD22 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Llama | |
| Human | |
| CAA42006.1 | |
| 933 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| L007.2.5LL | |
| Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Chinese Hamster Ovary cell line CHO-derived human Siglec-2/CD22, Asp20-Arg687, Accession # CAA42006.1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title