Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-3/CD33 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1137N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1137N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1137N100 Supplier R&D Systems Supplier No. FAB1137N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry
Classification Monoclonal
Clone 6C5/2
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Gene Symbols CD33
Host Species Mouse
Immunogen Recombinant human Siglec-3/CD33 Extracellular domain
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 945
Test Specificity Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.