Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-3/CD33 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Siglec-3/CD33 |
---|---|
Clone | 996810 |
Dilution | Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11371SP
![]() |
R&D Systems
MAB11371SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11371100
![]() |
R&D Systems
MAB11371100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
Siglec-3/CD33 | |
Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
Mouse myeloma cell line, NS0-derived human Siglec-3/CD33, Asp18-His259, Accession # P20138 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Siglec-3/CD33 in direct ELISAs. |
996810 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Human | |
P20138 | |
945 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title