Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-3/CD33 LlamabodyTM VHH His-tag Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. LMAB1073310
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LMAB1073310 100 μg
LMAB10733SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. LMAB1073310 Supplier R&D Systems Supplier No. LMAB10733100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Llama Monoclonal Antibody

Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Siglec-3/CD33
Applications Flow Cytometry
Classification Monoclonal
Clone L008.2.14N
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/106 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. AAA51948
Gene Alias CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Host Species Llama
Immunogen Mouse myeloma cell line NS0-derived human Siglec-3/CD33, Val18-His259, Accession # AAA51948
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 945
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.