Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-3/CD33 (Research Grade Gemtuzumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
$214.00 - $549.50
Specifications
Antigen | Siglec-3/CD33 |
---|---|
Clone | Hu9a |
Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
Applications | CyTOF, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9635SP
![]() |
R&D Systems
MAB9635SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9635100
![]() |
R&D Systems
MAB9635100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Siglec-3/CD33 | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
Human Siglec-3/CD33 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Siglec-3/CD33 based on Gemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Gemtuzumab. This product is for research use only. |
Hu9a | |
CyTOF, Flow Cytometry | |
Unconjugated | |
Human | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
945 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title