Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-5/Siglec-14 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems MAB10721
This item is not returnable.
View return policy
Description
Siglec-5/Siglec-14 Monoclonal specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
Siglec-5/Siglec-14 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.04-0.2 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
O15389 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8778 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Flow Cytometry, Neutralization, CyTOF, ELISA | |
194128 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SIGLEC5 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-5/Siglec-14 Lys18-Thr434 Accession # O15389 | |
500 μg | |
Primary | |
Detects human Siglec-5/Siglec-14 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant human (rh) Siglec-14 is observed and no cross-reactivity with rhSiglec-3, rhSiglec-7, or rhSiglec-9 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction