Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Siglec-5/Siglec-14 DuoSet ELISA
SDP

Catalog No. DY1072 Shop All R&D Systems Products
Change view
Click to view available options
Conjugate:
Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
DY1072 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY1072 Supplier R&D Systems™ Supplier No. DY1072
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Human Siglec-5/Siglec-14 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Siglec-5/Siglec-14.

Specifications

Conjugate Biotin
Assay Range 46.90 to 3000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 8778
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.