Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-6/CD327 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA2859RV100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA2859RV100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA2859RV100 Supplier R&D Systems Supplier No. FAB2859RV100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-6/CD327 Monoclonal specifically detects Siglec-6/CD327 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-6/CD327
Applications Flow Cytometry
Classification Monoclonal
Clone 767329R
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/10^6 cells
Formulation Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O43699
Gene Alias CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327 Gln27-Ser331 Accession # O43699
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 946
Test Specificity Detects human Siglec-6/CD327 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.