Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-6/CD327 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2859S100UG
This item is not returnable.
View return policy
Description
Siglec-6/CD327 Monoclonal specifically detects Siglec-6/CD327 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-6/CD327 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O43699 | |
SIGLEC6 | |
Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327 Gln27-Ser331 Accession # O43699 | |
RUO | |
946 | |
Human | |
IgG2a |
Flow Cytometry | |
767329 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6 | |
Mouse | |
100 μg | |
Primary | |
Detects human Siglec-6/CD327 in ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction