Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-7/CD328 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1138
This item is not returnable.
View return policy
Description
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot.Specifications
Siglec-7/CD328 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Y286 | |
SIGLEC7 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328 Gln19-Gly357 (predicted) Accession # Q9Y286 | |
500 μg | |
Primary | |
Detects human Siglec-7/CD328 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, or -9 is observed. | |
Human | |
Purified |
Western Blot | |
194212 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27036 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction