Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-7/CD328 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1138
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1138 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1138 Supplier R&D Systems Supplier No. BAF1138
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Siglec-7/CD328 Polyclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Siglec-7/CD328
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. Q9Y286
Gene Alias Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7
Gene Symbols SIGLEC7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328 Gln19-Gly357 Accession # Q9Y286
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27036
Test Specificity Detects human Siglec-7/CD328. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Siglec-9 is observed, less than 5% cross-reactivity with rhSiglec-3 is observed, and less than 1% cross-reactivity with rhSiglec-2 and rhSiglec-5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.