Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Human Siglec-8 Antibody, R&D Systems™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7975SP
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Siglec-8 Monoclonal specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Siglec-8 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9NYZ4 | |
| SIGLEC8 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-8 Met17-Ala363 Accession # Q9NYZ4 | |
| 25 μg | |
| Primary | |
| Detects human Siglec-8 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-7, -9, or -14 is observed. | |
| Human | |
| Purified | 
| Flow Cytometry, CyTOF | |
| 837535 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD329 antigen, CDw329, MGC59785, SAF2, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec8, Siglec-8, SIGLEC8L, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27181 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction