Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-9 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Catalog No. FAB1139U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1139U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1139U100 Supplier R&D Systems Supplier No. FAB1139U100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-9
Applications Flow Cytometry
Classification Monoclonal
Clone 191240
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Test Specificity Detects human Siglec-9 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Siglec-E is observed and less than 1% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5, rhSiglec-6, rhSiglec-7, rhSiglec-8 or rhSiglec-10 is observed. In Western blots, approximately 100% cross-reactivity with recombinant mouse Siglec-E is observed under non-reduced conditions.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.