Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-9 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$187.00 - $491.00
Specifications
Antigen | Siglec-9 |
---|---|
Clone | 191240 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1139G025
![]() |
R&D Systems
FAB1139G025 |
25 Tests |
Each for $187.00
|
|
|||||
FAB1139G100
![]() |
R&D Systems
FAB1139G100 |
100 Tests |
Each for $491.00
|
|
|||||
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-9 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
SIGLEC9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed. |
191240 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Q9Y336 | |
27180 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title