Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-9 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1139T100UG
This item is not returnable.
View return policy
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-9 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y336 | |
SIGLEC9 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
RUO | |
27180 | |
Human | |
IgG2a |
Flow Cytometry | |
191240 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
Mouse | |
100 μg | |
Primary | |
Detects human Siglec-9 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Siglec-E is observed and less than 1% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5, rhSiglec-6, rhSiglec-7, rhSiglec-8 or rhSiglec-10 is observed. In Western blots, approximately 100% cross-reactivity with recombinant mouse Siglec-E is observed under non-reduced conditions. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction