Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1139SP
This item is not returnable.
View return policy
Description
Siglec-9 Polyclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| Siglec-9 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27180 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| Q9Y336 | |
| SIGLEC9 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 Accession # Q9Y336 | |
| 25 μg | |
| Primary | |
| Detects human Siglec-9 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Siglec-7 is observed, 5% cross-reactivity with rhSiglec-3 and rhSiglec-5 is observed, and less than 1% cross-reactivity with rhSiglec-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction