Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Siglec-9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1139500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1139500 500 μg
MAB1139SP 25 μg
MAB1139100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1139500 Supplier R&D Systems Supplier No. MAB1139500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Siglec-9
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF, ELISA
Classification Monoclonal
Clone 191240
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.015-0.075 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Test Specificity Detects human Siglec-9 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Siglec-E is observed and less than 1% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5, rhSiglec-6, rhSiglec-7, rhSiglec-8 or rhSiglec-10 is observed. In Western blots, approximately 100% cross-reactivity with recombinant mouse Siglec-E is observed under non-reduced conditions.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.